Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multi-element standards, common and transition elements, VHG

Multi-element standards, common and transition elements, VHG

Common and transition element standards, 100 ml, 100 mL
  PROOVHG-SM75B-100
Multi-element standards, common and transition elements, VHG
  PROOVHG-SM75B-100
 :  
Multi-element standards, common and transition elements, VHG

Common and transition element standards, 100 ml, 100 mL

 :  
 
698,00 €
   
1 * 100 mL
 

This standard solution is intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).

Multi elements standards for Ag, Al, As, Ba, Be, Bi, Cd, Cr, Co, Cu, Fe, Li, Mn, Mo, Ni, Pb, Sb, Se, Sr, Tl, V, Zn at 100 µg/ml and Ca, K, Mg, Na at 1000 µg/ml in 0,2% HF.

Certifications: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.

 

 

  • Pk:
    100 mL
  • Description:
    Common and transition element standards
  • Pk Info:
    100 ml

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.