Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

Buffer solution, 1,00, ±0,02, 25 °C, Plastic bottle, 500 mL
Catalog # REGE1010525
Supplier:   REAGECON
Buffer solutions
Catalog # REGE1010525
Supplier:  REAGECON
CAS Number:  
Buffer solutions
Buffer solutions

Buffer solution, 1,00, ±0,02, 25 °C, Plastic bottle, 500 mL

CAS Number:  
Price
38,40 €
Log in for availability
1 * 500 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,02
  • pH value:
    1,00

Specifications