Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Electrolyte solutions

Electrolyte solution (KCl 3M), 250 ml, 250 mL
  HACHLZW9500.99
 :   Hach
Electrolyte solutions
  HACHLZW9500.99
 :  Hach
 :  
Electrolyte solutions
Electrolyte solutions

Electrolyte solution (KCl 3M), 250 ml, 250 mL

 :  
 
29,20 €
   
1 * 250 mL
 

 

 

 

  • Pk:
    250 mL
  • Description:
    Electrolyte solution (KCl 3M), 250 ml