Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sampling spoons, LaboPlast

Sampling spoons, LaboPlast

170 mm, 10 ml
Catalog # 231-0514
Supplier:   BURKLE
Sampling spoons, LaboPlast
Catalog # 231-0514
Supplier:  BURKLE
Sampling spoons, LaboPlast
Sampling spoons, LaboPlast

170 mm, 10 ml

Price
68,00 €
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents

PS, white. These disposable spoons are ideal for sampling powders, granules and pastes.

  • Available with capacity of 2,5 ml (similar to teaspoon) and 10 ml (similar to tablespoon)
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to EU food and FDA guidelines
  • Individually packed
Spécifications
  • Capacity:
    10 ml
  • Length:
    170 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.