Sampling scoops, SteriPlast®
With lid, 150 ml, 218 mm, White
Catalog # 231-0522
Supplier: BURKLE
Jump to:
Product Details & Documents
PS, white. These scoops are ideal for the sampling of powder, granules and pastes. They are also available with a transparent lid to close it immediately after the sampling procedure to secure a sterile and contamination-free sample.
- Individually packed, gamma radiation sterilised
- Production, assembling and packaging acc. to ISO cleanroom class 7
- According to EU food and FDA guidelines
Due to the special handle construction and the slightly angled edge the scoop stands horizontal and can be laid down without the material trickling out.
Documents
Specifications
- Type:With lid
- Capacity:150 ml
- Colour:White
- Length:218 mm
- Seq:08
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



