[EN]BUDESONIDE FOR SYSTEM SUITABILITY 1 * 10 mg
Catalog # EDQMY0001148
Supplier: EDQM
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo