Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

pH buffer solutions, NIST/DIN

NIST/DIN buffer solution, 30×20 ml, 9,180, 1
Catalog # METT30111138
Supplier:   Mettler - Toledo
pH buffer solutions, NIST/DIN
Catalog # METT30111138
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions, NIST/DIN

NIST/DIN buffer solution, 30×20 ml, 9,180, 1

CAS Number:  
Price
137,00 €
Log in for availability
1 * 30 ST
Product Details & Documents

The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision.

A detailed test certificate is available for every bottle and guarantees the tested values and traceability.

 : NIST traceable.

Specifications

Specifications

  • Pk:
    1
  • Description:
    NIST/DIN buffer solution, 30×20 ml
  • pH value:
    9,180

Specifications