Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions, NIST/DIN

pH buffer solutions, NIST/DIN

NIST/DIN buffer solution, 30×20 ml, 6,865, 1
N° de catalogue METT30111137
Fournisseur:   Mettler - Toledo
pH buffer solutions, NIST/DIN
N° de catalogue METT30111137
Fournisseur:  Mettler - Toledo
Numéro CAS:  
pH buffer solutions, NIST/DIN

NIST/DIN buffer solution, 30×20 ml, 6,865, 1

Numéro CAS:  
Prix
136,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision.

A detailed test certificate is available for every bottle and guarantees the tested values and traceability.

Certifications: NIST traceable.

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pk:
    1
  • Description:
    NIST/DIN buffer solution, 30×20 ml
  • pH value:
    6,865

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.