Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions, NIST/DIN

pH buffer solutions, NIST/DIN

NIST/DIN buffer solution, 30×20 ml, 4,006, 1
Catalog # METT30111136
Supplier:   Mettler - Toledo
pH buffer solutions, NIST/DIN
Catalog # METT30111136
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions, NIST/DIN

NIST/DIN buffer solution, 30×20 ml, 4,006, 1

CAS Number:  
Price
136,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision.

A detailed test certificate is available for every bottle and guarantees the tested values and traceability.

Certifications: NIST traceable.

Documents

Spécifications

Spécifications

  • Pk:
    1
  • Description:
    NIST/DIN buffer solution, 30×20 ml
  • pH value:
    4,006

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.