Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bath fluids, Haake

Bath fluids, Haake

Synth 200, +20...+210 °C, >235 °C, 227 °C, 100 cP, Light brown, 1
Catalog # HAKA9990225
Supplier:   THERMO ELECTRON LED
Bath fluids, Haake
Catalog # HAKA9990225
CAS Number:  
Bath fluids, Haake

Synth 200, +20...+210 °C, >235 °C, 227 °C, 100 cP, Light brown, 1

CAS Number:  
Price
582,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These carefully selected and proven heat transfer liquids have been designed to reduce health hazards and minimise unpleasant odours.

  • Silicone oils carry a low risk of inflammation, do not give off unpleasant odours and have a long service life
  • Low corrosive tendency

The oils are not suitable for contact with silicone tubing.

Documents

Specifications

Specifications

  • Pk:
    1
  • Colour:
    Light brown
  • Fire point:
    >235 °C
  • Flash point:
    227 °C
  • Temperature range:
    +20...+210 °C
  • Type:
    Synth 200
  • Viscosity:
    100 cP

Specifications