CUVETTE SYSTEM REACTION COD 5000-9 1 * 25 items
Catalog # WTWA252093
Supplier: WTW
Frequently Bought Together
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for UHPLC-MS
Supplier: VWR Chemicals
Methyl ethyl ketone ACS
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2,2,4-Trimethylpentane ≥98%, TECHNICAL
Supplier: VWR Chemicals
1-Propanol ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Benzyl alcohol, extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
tert-Butanol ACS
Supplier: Thermo Fisher Scientific
n-Heptane ≥99.3%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
2,2,4-Trimethylpentane ≥99.5%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone 99.9% (by GC, corrected for water content), ECD tested halocarbons free grade for residue analysis
Supplier: Thermo Fisher Scientific
Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific