BORIC ACID SOLUTION 4% W / V (CUSTOMIZED 1 * 5 L
FFCHFF64245000
Frequently Bought Together
Potassium iodide ≥99%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Sodium chloride ≥98%, TECHNICAL
: VWR Chemicals
di-Potassium hydrogen phosphate ≥99.0% analytical reagent
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals