Corning® Lymphocyte Separation Medium, Corning
LSM (Lymphocyte Separation Medium), 1×100 ml
Supplier: Corning
Jump to:
Product Details & Documents
Lymphocyte Separation Medium (LSM) was originally designed for the in vitro isolation of lymphocytes from diluted whole blood
It is a sterile, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/mL at 20ºC.
Specifications
- Description:LSM (Lymphocyte Separation Medium), 1×100 ml
- Seq:01