Volumetric overflow flasks, without stopper
94 ml, Overflow flask
Catalog # NEUB0372-10-094
Supplier: NEUBERT VOLUME GLASSWAERE
Volumetric overflow flasks, without stopper
Catalog # NEUB0372-10-094
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
These volumetric overflow flasks are made of borosilicate glass 3.3.
- Calibrated to deliver (TD, Ex)
- Cut and ground exactly at the volume-mark
- Without stopper
Specially designed for water analysis.
Specifications
- Capacity:94 ml
- Description:Overflow flask
- Seq:02