Loop cartridge, 100 μL, right hand side, for Agilent G7167 Multisampler 1 * 1 items
Catalog # HEWLG4267-60500
Supplier: Agilent
Loop cartridge, 100 μL, right hand side, for Agilent G7167 Multisampler 1 * 1 items
Catalog # HEWLG4267-60500
Supplier: Agilent
Frequently Bought Together
Caps for 4 ml glass screw top vials, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
SafetyWasteCaps
Supplier: S.C.A.T.
Capillary column ferrules for gas chromatography
Supplier: Agilent
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Snap Cap Vials
Supplier: VWR Collection
Liquid-liquid extraction, EXtrelut® NT
Supplier: Merck
Silica gels for preparative chromatography, standardised
Supplier: MERCK PRODUCTION CHEMICALS
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
HPLC columns, NUCLEOSIL® C18
Supplier: MACHEREY-NAGEL
Headspace crimp vials, ND20, La-Pha-Pack™
Supplier: LA PHA PACK
Liquid chromatography sorbents, Polyamide 6
Supplier: MACHEREY-NAGEL
Supplies for pumps
Supplier: Agilent
J.T.Baker® BAKERBOND spe™ C18, SPE Columns
Supplier: Avantor
Adapter for exhaust filter for Justrite and CPC canisters
Supplier: S.C.A.T.
Screw caps for screw neck vials, N 9
Supplier: MACHEREY-NAGEL
Calibration standard, single element, silicon for AAS and ICP-OES, pure grade
Supplier: PerkinElmer