Orders LinkContactUsLinkComponent
Spray chambers and brackets
Spray chambers and brackets
UniFit drain connector, 2,0 mm ext. Ø, 0,86 mm int. Ø, for spray chamber used with 5100 dual view ICP-OES, with 480 mm capillary tubing
Catalog # HEWLG8010-80036
Supplier:   Agilent
Spray chambers and brackets
Catalog # HEWLG8010-80036
Supplier:  Agilent
Spray chambers and brackets
Spray chambers and brackets
UniFit drain connector, 2,0 mm ext. Ø, 0,86 mm int. Ø, for spray chamber used with 5100 dual view ICP-OES, with 480 mm capillary tubing
Price
108,00 €
Log in for availability
1 * 3 ST
Product Details & Documents
These ICP-OES series are compatible with a range of accessories and supplies designed to extend the capabilities of the instruments.
Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout.
Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Sturman-Masters spray chamber is inert and minimizes interferences and improves stability with high TDS samples.
Specifications
  • Description:
    UniFit drain connector, 2,0 mm ext. Ø, 0,86 mm int. Ø, for spray chamber used with 5100 dual view ICP-OES, with 480 mm capillary tubing
  • Seq:
    07