Syringes, 3-piece
Without needle, Luer eccentric tip, 50 ml
Supplier: TERUMO
Jump to:
Product Details & Documents
Sterile, disposable, three-piece, latex-free.
- Available with Luer slip tip or Luer lock threaded tip
- Sterilised by electron beam irradiation
Certifications: Compliant with Directive 93/42/EEC Class IIa, organisation accredited by TÜV Rheinland (Germany), factory certified to ISO 9001 EN 46001, sterilisation process validated according to Standard NF EN 550.
Packaging: Individual sterile blister packs.
Specifications
- Capacity:50 ml
- Description:Without needle, Luer eccentric tip
- Seq:20