ContactUsLinkComponent
Bell-flo, complete set, 1L, bottom indents and three vertical baffles for optimal mixing, 32mm or 45mm sidearms for port assemblies, rugged stainless steel/PTFE internal bearing assemblies. 1 * 1 items
Catalog # BELC1965-81005
Supplier:  BELLCO GLASS
Bell-flo, complete set, 1L, bottom indents and three vertical baffles for optimal mixing, 32mm or 45mm sidearms for port assemblies, rugged stainless steel/PTFE internal bearing assemblies. 1 * 1 items
Catalog # BELC1965-81005
Supplier:  BELLCO GLASS
Frequently Bought Together