Orders LinkContactUsLinkComponent
Test kits, drop count tests
Test kits, drop count tests
Formaldehyde, model FM-1, 0,5 - 10%, 1 KIT
Catalog # HACH2183100
Supplier:   Hach
Test kits, drop count tests
Catalog # HACH2183100
Supplier:  Hach
CAS Number:  
Test kits, drop count tests
Test kits, drop count tests
Formaldehyde, model FM-1, 0,5 - 10%, 1 KIT
CAS Number:  
Price
184,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Formaldehyde, model FM-1
  • Range:
    0,5 - 10%
Specifications