Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Conductivity standard solution, 147 µS/cm, 250 mL
Catalog # HACHLZW9700.99
Supplier:   Hach
Conductivity standards
Catalog # HACHLZW9700.99
Supplier:  Hach
CAS Number:  
Conductivity standards
Conductivity standards
Conductivity standard solution, 147 µS/cm, 250 mL
CAS Number:  
Price
25,20 €
Log in for availability
1 * 250 mL
Product Details & Documents
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Specifications
Specifications
  • Pk:
    250 mL
  • Description:
    Conductivity standard solution
  • Conductivity:
    147 µS/cm
Specifications