Order Entry
Belgium
Orders LinkContactUsLinkComponent
Halogen bulbs

Halogen bulbs

Quartz halogen bulb, 12 V, 100 W
Catalog # 630-2568
Supplier:   MOTIC
Halogen bulbs
Catalog # 630-2568
Supplier:  MOTIC
Halogen bulbs
Halogen bulbs

Quartz halogen bulb, 12 V, 100 W

Price
24,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Quartz halogen bulb
  • Seq:
    05
  • Power:
    100 W
  • Voltage:
    12 V
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.