Orders LinkContactUsLinkComponent
Trays
Trays
Flask tray with threaded holes to attach 25 to 1000 ml SC flask clamps, SH plate holders and SR racks, for LSB12
Catalog # 462-0537
Supplier:   GRANT INSTRUMENTS
Trays
Catalog # 462-0537
Supplier:  GRANT INSTRUMENTS
Trays
Trays
Flask tray with threaded holes to attach 25 to 1000 ml SC flask clamps, SH plate holders and SR racks, for LSB12
Price
329,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Flask tray with threaded holes to attach 25 to 1000 ml SC flask clamps, SH plate holders and SR racks, for LSB12
  • Seq:
    01