Orders LinkContactUsLinkComponent
Trays
Trays
Stainless steel perforated base tray, allows bath to be used as an unstirred bath, for LSB12
Catalog # 462-0520
Supplier:   GRANT INSTRUMENTS
Trays
Catalog # 462-0520
Supplier:  GRANT INSTRUMENTS
Trays
Trays
Stainless steel perforated base tray, allows bath to be used as an unstirred bath, for LSB12
Price
138,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Stainless steel perforated base tray, allows bath to be used as an unstirred bath, for LSB12
  • Seq:
    10