Orders LinkContactUsLinkComponent
Caps, closed top
Caps, closed top
With bonded PTFE Liner, bulk, 38 - 400 mm
Catalog # EPSCB167-3800BL
   Low Manufacturing Impact
Supplier:   EP SCIENTIFIC
Caps, closed top
Catalog # EPSCB167-3800BL
Supplier:  EP SCIENTIFIC
   Low Manufacturing Impact
Caps, closed top
Caps, closed top
With bonded PTFE Liner, bulk, 38 - 400 mm
Price
33,90 €
Connexion Pour la disponibilité
1 * 24 ST
Product Details & Documents
PP, white.
  • Very good chemical resistance
  • Very good temperature resistance
Unprocessed caps are ideal for environmental sample bottles, jars, jugs and vials.
Certifications: Manufactured under an ISO certified quality management system
Spécifications
  • Cap Size:
    38 - 400 mm
  • Description:
    With bonded PTFE Liner, bulk
  • Seq:
    04
  • Cat. No.:
    EPSCB167-3800BL
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact