Screw caps
Caps, PP, white, LDPE foam lined, 38-400
EPSC067-3800WBP
LDPE-lined white caps.
- For general-purpose HDPE bulk separate containers
- Same high quality caps used on certified HDPE environmental sample containers
: Bottles sold separately (IR013-series and IR012-series).
- Thread:38-400
- Description:Caps, PP, white, LDPE foam lined
- Seq:03
- Cat. No.:EPSC067-3800WBP
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact