Orders LinkContactUsLinkComponent
Disposable filters, Optimizer®
Disposable filters, Optimizer®
D disposable filter with 6,35 mm connection, 6,35 mm vent/drain compression seal fittings, Ø×L: 69×113 mm, 0,02 µm, 2200 cm²
Catalog # ENTECWMX0S1S3
Supplier:   ENTEGRIS
Disposable filters, Optimizer®
Catalog # ENTECWMX0S1S3
Supplier:  ENTEGRIS
Disposable filters, Optimizer®
D disposable filter with 6,35 mm connection, 6,35 mm vent/drain compression seal fittings, Ø×L: 69×113 mm, 0,02 µm, 2200 cm²
Price
Inquire For Price
Log in for availability
1 * 1 SET
Product Details & Documents
Photochemical filter with superior contaminant retention and low hold-up volume performance in sub-10 to 180 nm technology nodes.
Specifications
  • Filtration area:
    2200 cm²
  • Pore size:
    0,02 µm
  • Type:
    D disposable filter with 6,35 mm connection, 6,35 mm vent/drain compression seal fittings, Ø×L: 69×113 mm
  • Seq:
    04