ContactUsLinkComponent
C18G x 3µm particle size x 300Å pore size, 10mm length x 4.0mm ID, Stainless steel PK3 ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2C184-03G04S
C18G x 3µm particle size x 300Å pore size, 10mm length x 4.0mm ID, Stainless steel PK3 ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2C184-03G04S
Frequently Bought Together