ContactUsLinkComponent
C18G x 3µm particle size x 200Å pore size, 10mm length x 2.1mm ID, Stainless steel PK3 ProteCol HPLC Guard Column PK3 1 * 1 items
  SGEA2C181-03G02S
C18G x 3µm particle size x 200Å pore size, 10mm length x 2.1mm ID, Stainless steel PK3 ProteCol HPLC Guard Column PK3 1 * 1 items
  SGEA2C181-03G02S
Frequently Bought Together