Order Entry
Belgium
Orders LinkContactUsLinkComponent
Counting chambers, C-Chip

Counting chambers, C-Chip

Counting chambers, Malassez, 0,2 mm, 25×75 mm
Catalog # 631-1097
Supplier:   NANOENTEK
Counting chambers, C-Chip
Catalog # 631-1097
Supplier:  NANOENTEK
Counting chambers, C-Chip
Counting chambers, C-Chip

Counting chambers, Malassez, 0,2 mm, 25×75 mm

Price
272,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

Quartz grade optical plastic counting chambers applicable in blood analysis (hematology).

  • Guaranteed repeatability and reliability, eliminate the need for washing and reuse
  • Disposable hemocytometers that do not need coverslips
  • Chip to reduce exposure to infectious samples and contamination

Suitable for cell counting and culture, concentration measurement and viability test, in bacteria and fungal spore counting, IVF, IUI tasks of sperm counting.

Specifications
  • Depth of chamber:
    0,2 mm
  • Type:
    Malassez
  • W×D:
    25×75 mm
  • Description:
    Counting chambers
  • Seq:
    07