ContactUsLinkComponent
Clearklens Ipa Sterile 1L, 70% Ipa With 30% Wfi, Filtered To 0,2µm. Aseptically Filled In 1L Capped Bottles, Double-Bagged. With Certificate Of Sterility. 1 * 10 l
Catalog # DIVY7522373_U
Supplier:  Diversey
Clearklens Ipa Sterile 1L, 70% Ipa With 30% Wfi, Filtered To 0,2µm. Aseptically Filled In 1L Capped Bottles, Double-Bagged. With Certificate Of Sterility. 1 * 10 l
Catalog # DIVY7522373_U
Supplier:  Diversey
Frequently Bought Together