Ion selective electrodes, Fluoride, IntelliCAL™
Product Details & Documents
IntelliCAL™ ISEF121 is a digital combination fluoride ion selective electrode (ISE) with a non-refillable, single junction reference and built-in temperature sensor. The solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
- Compatible with digital Hach meters only (e.g. HQd meters)
- Requires virtually no maintenance
- Fast, stable, and accurate results
- Ultimate traceability in measurement history
- Can be moved between meters without the need to re-calibrate or re-enter measurement settings
The probe is available with a 1 or 3 meter cable and is intended for laboratory use. Ideal for measuring fluoride concentrations in drinking water, wastewater, and general water quality applications.
Ordering information: Electrode is supplied with integrated temperature sensor, calibration certificate, and basic user manual.
The analysis package contains the IntelliCAL™ ISEF12101 electrode, fluoride ionic strength adjustor powder pillows (100), fluoride 0,5 mg/L standard solution (500 ml), fluoride 1,0 mg/L standard solution (500 ml), and fluoride 2,0 mg/L standard solution (500 ml).
Documents
Specifications
- Concentration:0,01 - 19000 mg/l F-
- Connection:1 m cable
- Ø×L:12×220 mm
- Type:IntelliCAL™ ISEF121 (Fluoride)
- Description:IntelliCAL™ fluoride electrode ISEF121, Ag/AgCl reference system
- Material:Epoxy
- Seq:50
- Diaphr.:Porous PTFE annular ring
- Electrolyte:driTEK gel (non refillable)
- Temp.:5...50 °C
Accessories
Product Information
- Catalog #TypePkAvailabilityPrice
- Fluoride ionic strength adjustor (ISA) powder pillows, 0,01 mg/L to saturatedFluoride ionic strength adjustor (ISA) powder pillows, 0,01 mg/L to saturatedHACH258999PP100Log in for availability1 * 100 ST - 132,00 €
Product Information
- Catalog #ConcentrationAccuracyAvailabilityPrice
- Fluoride standard solutionFluoride standard solutionHACH291491,00 µg/ml±0,02 µg/mlLog in for availability1 * 500 mL - 40,20 €



