Order Entry
Belgium
Orders LinkContactUsLinkComponent
Retort rods, without thread

Retort rods, without thread

12 mm, 230 mm
Catalog # BOCH5120-230
Supplier:   BOCHEM
Retort rods, without thread
Catalog # BOCH5120-230
Supplier:  BOCHEM
Retort rods, without thread
Retort rods, without thread

12 mm, 230 mm

Price
21,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10.

  • Rods for stand bases
  • Without thread
Specifications
  • Ø:
    12 mm
  • Length:
    230 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.