ContactUsLinkComponent
Splitter kit, 1 to 5 mL/min, analytical, for evaporative light scattering detector. Includes capillary to ELSD, 0.18 mm, 330 mm (502 3-2260), capillary to fraction collector-autosampler, 0.25 mm, 220 mm (5023-2261), high-flow union, stainless steel, 1 * 1 items
Catalog # HEWL5023-2255
Supplier:  Agilent
Splitter kit, 1 to 5 mL/min, analytical, for evaporative light scattering detector. Includes capillary to ELSD, 0.18 mm, 330 mm (502 3-2260), capillary to fraction collector-autosampler, 0.25 mm, 220 mm (5023-2261), high-flow union, stainless steel, 1 * 1 items
Catalog # HEWL5023-2255
Supplier:  Agilent
Frequently Bought Together