ContactUsLinkComponent
Splitter kit, 2 to 10 mL/min, analytical, for Agilent G4260B evaporative light scattering detector. Includes capillary to ELSD, 0.18 mm, 330 mm (5023-2260), capillary to fraction collector, 0.25 mm, 500 mm (5023-2262), high-flow union, stainless ste 1 * 1 items
Catalog # HEWL5023-2256
Supplier:  Agilent
Splitter kit, 2 to 10 mL/min, analytical, for Agilent G4260B evaporative light scattering detector. Includes capillary to ELSD, 0.18 mm, 330 mm (5023-2260), capillary to fraction collector, 0.25 mm, 500 mm (5023-2262), high-flow union, stainless ste 1 * 1 items
Catalog # HEWL5023-2256
Supplier:  Agilent
Frequently Bought Together