ContactUsLinkComponent
Splitter kit, 25 to 100 mL/min, preparative, for Agilent G4260B evaporative light scattering detector. Includes capillary to ELSD, 0 .18 mm, 300 mm (5023-2260), capillary to fraction collector, 0.75 mm, 600 mm (5023-2266), tubing, PTFE, 0.7 mm id, 1. 1 * 1 items
  HEWL5023-2258
 :  Agilent
Splitter kit, 25 to 100 mL/min, preparative, for Agilent G4260B evaporative light scattering detector. Includes capillary to ELSD, 0 .18 mm, 300 mm (5023-2260), capillary to fraction collector, 0.75 mm, 600 mm (5023-2266), tubing, PTFE, 0.7 mm id, 1. 1 * 1 items
  HEWL5023-2258
 :  Agilent
Frequently Bought Together