ContactUsLinkComponent
qPCR detection of Rotavirus C. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses onl y), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-ROTAC
Supplier:  PRIMERDESIGN
qPCR detection of Rotavirus C. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses onl y), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-ROTAC
Supplier:  PRIMERDESIGN
Frequently Bought Together