Orders LinkContactUsLinkComponent
Capillary tubes
Capillary tubes
Sealed both ends, 100 mm, 1,8/2,0 mm
Catalog # 621-0050
Supplier:   MURRAY S
Capillary tubes
Catalog # 621-0050
Supplier:  MURRAY S
Capillary tubes
Capillary tubes
Sealed both ends, 100 mm, 1,8/2,0 mm
Price
191,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Soda glass.
  • For melting point determinations
Specifications
  • Ext.Ø:
    1,8/2,0 mm
  • Description:
    Sealed both ends
  • Length:
    100 mm