Detectable plasters
Detectable finger plasters, blue, refill set for dispenser, 10 plasters, 180×20 mm, 160×57×122 mm
SOEH1009988
: SOEHNGEN
These blue, detectable plasters are specially designed for use in the food industry. Dispenser containing aluminium strips with an integrated metal surface that can be detected via a metal detector.
- Plasters are detectable by most metal detectors used within the food industry
- Breathable and skin-friendly
- High quality hypoallergenic plasters
- Description:Detectable finger plasters, blue, refill set for dispenser, 10 plasters
- L×W:180×20 mm
- Seq:04
- W×D×H:160×57×122 mm



