ContactUsLinkComponent
FireDebris tubular rollstock offers superior fire evidence collection and retention. This 457 mm wide tubular rollstock is made of nylon and hydrocarbon free. Since it can be cut to any length, it allows for evidence of many different sizes to be col 1 * 1 Roll
Catalog # AMPATRS-18125-F
Supplier:  ProAmpac
undefined
FireDebris tubular rollstock offers superior fire evidence collection and retention. This 457 mm wide tubular rollstock is made of nylon and hydrocarbon free. Since it can be cut to any length, it allows for evidence of many different sizes to be col 1 * 1 Roll
Catalog # AMPATRS-18125-F
Supplier:  ProAmpac
Frequently Bought Together