Closures for storage vials
PFA, plain threaded, Micro columns, tubes and vials, 33 mm
Catalog # SAVI600-033-01
Supplier: SAVILLEX
Jump to:
Product Details & Documents
Specifications
- Cap Size:33 mm
- For:Micro columns, tubes and vials
- Type:PFA, plain threaded