Storage vials
Standard vial with round interior and round exterior, 7,00 ml, 22,1×37,1 mm, No, 24 mm
Catalog # SAVI200-007-20
Supplier: SAVILLEX
Jump to:
Product Details & Documents
PFA vials with round or conical interior.
- Ideal for storage and digestion in geochemistry
- Flat base for maximum heating efficiency
- Square exterior vials with hex closures have higher pressure rating
: Closures can be ordered separately. Modifications may be available on request.
Specifications
- Cap included:No
- Cap Size:24 mm
- Ø×H:22,1×37,1 mm
- Capacity:7,00 ml
- Description:Standard vial with round interior and round exterior



