ContactUsLinkComponent
Dialysis tubing Trial kit Spectra/Por 7 pre-treated standard regenerated cellulose (RC) membrane MWCO 1kD 38mm flat width (24mm diam eter) 4,6ml/cm including tubing closures and openers wet 1 * 1 m
Catalog # SPEC132104T
Dialysis tubing Trial kit Spectra/Por 7 pre-treated standard regenerated cellulose (RC) membrane MWCO 1kD 38mm flat width (24mm diam eter) 4,6ml/cm including tubing closures and openers wet 1 * 1 m
Catalog # SPEC132104T
Frequently Bought Together