ContactUsLinkComponent
Transparent bag designed as primary and secondary packaging combining traceability and unique security to prevent fraud attempts while preserving content integrity during transfers and transport of samples. High security lock, unique sequential track 1 * 1.000 items
Catalog # 129-1034
Supplier:  COVERIS
Transparent bag designed as primary and secondary packaging combining traceability and unique security to prevent fraud attempts while preserving content integrity during transfers and transport of samples. High security lock, unique sequential track 1 * 1.000 items
Catalog # 129-1034
Supplier:  COVERIS
Frequently Bought Together