Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH Buffers

pH Buffers

pH buffer, 3,9 M NaCl, 7,00, 6×250 ml, 6
Catalog # METN52004107
Supplier:   Mettler - Toledo
pH Buffers
Catalog # METN52004107
Supplier:  Mettler - Toledo
CAS Number:  
pH Buffers

pH buffer, 3,9 M NaCl, 7,00, 6×250 ml, 6

CAS Number:  
Price
362,00 €
Log in for availability
1 * 6 ST
Product Details & Documents
Specifications

Specifications

  • Pk:
    6
  • Description:
    pH buffer, 3,9 M NaCl
  • Pk Info:
    6×250 ml
  • pH value:
    7,00

Specifications