Syringes, 3-piece
Without needle, Luer slip tip, 2,5 ml
Catalog # 613-3875
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
Sterile, disposable, three-piece, latex-free.
- Available with Luer slip tip or Luer lock threaded tip
- Sterilised by electron beam irradiation
: Compliant with Directive 93/42/EEC Class IIa, organisation accredited by TÜV Rheinland (Germany), factory certified to ISO 9001 EN 46001, sterilisation process validated according to Standard NF EN 550.
: Individual sterile blister packs.
Specifications
- Capacity:2,5 ml
- Description:Without needle, Luer slip tip
- Seq:03