Order Entry
Belgium
Orders LinkContactUsLinkComponent
Coloured cap inserts

Coloured cap inserts

Green
  525-0689
 :   Brand
Coloured cap inserts
  525-0689
 :  Brand
Coloured cap inserts
Coloured cap inserts

Green

 
21,80 €
   
1 * 500 ST
 

PP.

  • For quick identification
  • For microtubes 211-3201 to 211-3205 and 211-3211 to 211-3215
  • Applications: −196 to +121 °C
 
  • Colour:
    Green
  • Seq:
    02