0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
Orders Link
ContactUsLinkComponent
Cap Inserts
Coloured cap inserts
Coloured cap inserts
White
525-0690
:
Brand
Coloured cap inserts
525-0690
:
Brand
Coloured cap inserts
White
21,80 €
1 * 500 ST
-
+
PP.
For quick identification
For microtubes 211-3201 to 211-3205 and 211-3211 to 211-3215
Applications: −196 to +121 °C
Colour:
White
Seq:
04
Colour coders for cryovials, CryoColorCode, Nunc™
#: 479-2488
: Thermo Fisher Scientific
Combitips® advanced PCR Clean - Positive Displacement Pipette Tips
#: 613-2082
: EPPENDORF