Orders LinkContactUsLinkComponent
Accessories
Accessories
Microfilter fine black, PG 8536
Catalog # MIEL2154891
Accessories
Catalog # MIEL2154891
Supplier:  
Accessories
Accessories
Microfilter fine black, PG 8536
Discontinued
Product Details & Documents
  • For lower and upper baskets
  • With 7 x 7 mm perforations
  • Stainless steel
Specifications
  • Description:
    Microfilter fine black
  • Seq:
    23
  • For:
    PG 8536