3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use
1.000 mL
J63202.AU
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use
J63202.AU
: 54827-17-7
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result.
: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
- :54827-17-7
- :C₁₆H₂₀N₂
- :MFCD00007748
- :240.35 g/mol
- :Refrigerator
- :3,II
- Pk:1.000 mL