3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use
500 mL
Catalog # J63202.AP
Supplier: Thermo Fisher Scientific
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use
Catalog # J63202.AP
Supplier: Thermo Fisher Scientific
CAS Number: 54827-17-7
Jump to:
Product Details & Documents
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result.
Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
- CAS Number:54827-17-7
- Formula:C₁₆H₂₀N₂
- MDL Number:MFCD00007748
- Molecular Weight:240.35 g/mol
- Storage Temperature:Refrigerator
- ADR:3,II
Specifications
Specifications
- Pk:500 mL



